Search
Search

GCH1| Gene Symbol: GCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000161.2 | NP_000152.1 | 3, 4 | 675 | ||
| NM_001024024.1 | NP_001019195.1 | 3, 4 | 675 | |||
| NM_001024070.1 | NP_001019241.1 | 3, 4 | 675 | |||
| NM_001024071.1 | NP_001019242.1 | 3, 4 | 675 | |||
| XM_017021218.1 | XP_016876707.1 | 3, 4 | 1107 | |||
| GenBank mRNA | AY137463.1 | - | 3, 4 | 297 | ||
| AY137464.1 | - | 3, 4 | 297 | |||
| BC025415.2 | - | 3, 4 | 662 | 4129035 | ||
| CR536551.1 | - | 3, 4 | 514 | |||
| S43856.1 | - | 3, 4 | 581 | |||
| S44049.1 | - | 3, 4 | 488 | |||
| S44053.1 | - | 3, 4 | 581 | |||
| U19523.1 | - | 3, 4 | 662 | |||
| U66095.1 | - | 3, 4 | 658 | |||
| U66097.1 | - | 3, 4 | 667 | |||
| Z16418.1 | - | 3, 4 | 338 | |||
| Z29433.2 | - | 3, 4 | 613 | |||
| Z29434.2 | - | 3, 4 | 661 | |||