Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDE4DIP SIRNA Assays ›
          Gene Symbol:
          PDE4DIP LOC100996724
          Gene Name:
          phosphodiesterase 4D interacting protein
          phosphodiesterase 4D interacting protein-like
          Gene Aliases:
          CMYA2, MMGL
          -
          Chromosome Location:
          Chr.1: 148808167 - 149051277 on Build GRCh38
          Chromosome Location:
          Chr.1: 120471449 - 120701470 on Build GRCh38
          UniGene ID:
          Hs.584841
          Hs.731111
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s57314

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDE4DIP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198832.2 NP_001185761.2 28, 29 4017
          NM_001198834.3 NP_001185763.3 25, 26 4065
          NM_014644.5 NP_055459.5 25, 26 4065
          XM_005272981.3 XP_005273038.2 21, 22 4692
          XM_005277442.4 XP_005277499.2 21, 22 4692
          XM_005277443.3 XP_005277500.1 21, 22 4692
          XM_006711642.2 XP_006711705.1 21, 22 4692
          XM_006711644.2 XP_006711707.1 21, 22 4692
          XM_006711646.1 XP_006711709.1 28, 29 4354
          XM_006711648.2 XP_006711711.1 21, 22 4362
          XM_006711650.3 XP_006711713.1 26, 27 3839
          XM_006711651.1 XP_006711714.1 25, 26 3957
          XM_011510172.1 XP_011508474.1 28, 29 4512
          XM_011510174.1 XP_011508476.1 21, 22 4692
          XM_011510176.1 XP_011508478.1 28, 29 4182
          XM_017002875.1 XP_016858364.1 21, 22 4692
          XM_017002876.1 XP_016858365.1 21, 22 4692
          XM_017002878.1 XP_016858367.1 28, 29 4512
          XM_017002879.1 XP_016858368.1 21, 22 4692
          XM_017002880.1 XP_016858369.1 28, 29 4512
          XM_017002881.1 XP_016858370.1 21, 22 4692
          XM_017002883.1 XP_016858372.1 21, 22 4692
          XM_017002884.1 XP_016858373.1 21, 22 4692
          XM_017002885.1 XP_016858374.1 21, 22 4362
          XM_017002887.1 XP_016858376.1 29, 30 4789
          XM_017002888.1 XP_016858377.1 29, 30 4762
          XM_017002889.1 XP_016858378.1 29, 30 4434
          XM_017002890.1 XP_016858379.1 21, 22 4692
          XM_017002891.1 XP_016858380.1 27, 28 4593
          XM_017002892.1 XP_016858381.1 27, 28 4224
          XM_017002893.1 XP_016858382.1 28, 29 4712
          XM_017002894.1 XP_016858383.1 28, 29 4357
          XM_017002895.1 XP_016858384.1 25, 26 3675
          XM_017002896.1 XP_016858385.1 21, 22 4692
          XM_017002898.1 XP_016858387.1 28, 29 4354
          XM_017002899.1 XP_016858388.1 28, 29 4024
          XM_017002900.1 XP_016858389.1 21, 22 4362
          XM_017002901.1 XP_016858390.1 21, 22 4362
          XM_017002903.1 XP_016858392.1 28, 29 4024
          GenBank mRNA AB007923.2 - 29, 30 3896
          AL833273.1 - 25, 26 3953
          BX647660.1 - 25, 26 3993
          DA238288.1 - 2, 3 258
          Gene Symbol: LOC100996724
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017003064.1 XP_016858553.1 26, 27 3929
          GenBank mRNA CR749273.1 - 6, 7 1197

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein complex assembly
          protein binding
          enzyme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline