Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EHMT1 SIRNA Assays ›
          Gene Symbol:
          EHMT1
          Gene Name:
          euchromatic histone lysine methyltransferase 1
          Gene Aliases:
          EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
          Chromosome Location:
          Chr.9: 137618988 - 137836127 on Build GRCh38
          UniGene ID:
          Hs.495511
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s57320

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EHMT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_024757.4 NP_079033.4 24, 25 3491
          XM_005266105.4 XP_005266162.1 24, 25 3534
          XM_005266110.1 XP_005266167.1 23, 24 3406
          XM_006717288.2 XP_006717351.1 24, 25 3452
          XM_011519021.2 XP_011517323.1 24, 25 4489
          XM_011519022.2 XP_011517324.1 24, 25 3944
          XM_011519023.2 XP_011517325.1 24, 25 4468
          XM_011519024.2 XP_011517326.1 23, 24 4412
          XM_011519025.1 XP_011517327.1 24, 25 3654
          XM_011519026.2 XP_011517328.1 23, 24 4347
          XM_011519029.2 XP_011517331.1 16, 17 2079
          XM_011519030.2 XP_011517332.1 10, 11 1774
          XM_011519031.1 XP_011517333.1 10, 11 1132
          XM_011519033.2 XP_011517335.1 23, 24 4326
          XM_017015134.1 XP_016870623.1 24, 25 3922
          XM_017015135.1 XP_016870624.1 24, 25 3474
          XM_017015136.1 XP_016870625.1 23, 24 3396
          XM_017015137.1 XP_016870626.1 24, 25 3633
          XM_017015138.1 XP_016870627.1 23, 24 3385
          GenBank mRNA AB028932.1 - 19, 20 2311
          AB058779.2 - 19, 20 2311
          AF461894.1 - 1, 2 1178
          AL713772.1 - 2, 3 807
          AL832865.1 - 2, 3 807
          AY083210.1 - 22, 23 3361
          BC011608.2 - 14, 15 1708

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA methylation
          embryo development
          chromatin modification
          histone methylation
          peptidyl-lysine monomethylation
          peptidyl-lysine dimethylation
          negative regulation of transcription, DNA-templated
          histone H3-K9 methylation
          response to fungicide
          histone H3-K27 methylation
          regulation of signal transduction by p53 class mediator
          p53 binding
          protein binding
          methyltransferase activity
          zinc ion binding
          protein-lysine N-methyltransferase activity
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K9 specific)
          histone methyltransferase activity (H3-K27 specific)
          C2H2 zinc finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0