Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FCGR3B SIRNA Assays ›
          Gene Symbol:
          FCGR3B FCGR3A
          Gene Name:
          Fc fragment of IgG receptor IIIb
          Fc fragment of IgG receptor IIIa
          Gene Aliases:
          CD16, CD16b, FCG3, FCGR3, FCR-10, FCRIII, FCRIIIb
          CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
          Chromosome Location:
          Chr.1: 161623196 - 161631963 on Build GRCh38
          Chromosome Location:
          Chr.1: 161541759 - 161550623 on Build GRCh38
          UniGene ID:
          Hs.372679
          Hs.372679
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s57397

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FCGR3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000570.4 NP_000561.3 4 405
          NM_001244753.1 NP_001231682.1 3 651
          NM_001271035.1 NP_001257964.1 3 648
          NM_001271036.1 NP_001257965.1 3 517
          NM_001271037.1 NP_001257966.1 3 517
          GenBank mRNA AB025256.1 - 3 316
          AJ581669.1 - 3 316
          AK313219.1 - 3 397
          AK316565.1 - 3 337
          BC128562.1 - 3 300 40126686
          DA670720.1 - 3 517
          DA671001.1 - 3 502
          DA672751.1 - 4 405
          DA672763.1 - 3 557
          DA673839.1 - 3 414
          J04162.1 - 3 300
          M24854.1 - 3 381
          X07934.1 - 3 316
          X16863.1 - 3 316
          Gene Symbol: FCGR3A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127593.1 NP_001121065.1 4 438
          NM_001127595.1 NP_001121067.1 4 420
          NM_001127596.1 NP_001121068.1 4 417
          XM_011509293.2 XP_011507595.2 3 327
          GenBank mRNA AK223268.1 - 3 299
          AK223295.1 - 3 311
          AK302126.1 - 3 650
          BC017865.1 - 3 331 4690249
          BC033678.1 - 3 306 5180561
          BC036723.1 - 3 575 5206097
          BI768140.1 - 3 575
          DA674960.1 - 4 397
          DA933259.1 - 4 411
          DA944383.1 - 3 344
          DA957430.1 - 4 436
          KU178060.1 - 3 391
          KU178061.1 - 3 391
          M24853.1 - 1 193
          S76824.1 - 1 220
          X52645.1 - 3 316

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          immune response
          mitophagy in response to mitochondrial depolarization
          xenophagy
          Fc-gamma receptor signaling pathway involved in phagocytosis
          regulation of immune response
          IgG binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline