Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGAP4 SIRNA Assays ›
          Gene Symbol:
          AGAP4 AGAP12P AGAP5 AGAP7P AGAP9 LOC107984026
          Gene Name:
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 4
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 12, pseudogene
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 5
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 7, pseudogene
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 9
          arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
          Gene Aliases:
          AGAP-4, AGAP-8, AGAP8, CTGLF1, CTGLF5, MRIP2
          CTGLF12P
          CTGLF2
          AGAP-7, AGAP7, CTGLF4, bA109G10.1
          AGAP-9, CTGLF6, bA301J7.2
          -
          Chromosome Location:
          Chr.10: 45825592 - 45858529 on Build GRCh38
          Chromosome Location:
          Chr.10: 48010132 - 48031640 on Build GRCh38
          Chromosome Location:
          Chr.10: 73674275 - 73697796 on Build GRCh38
          Chromosome Location:
          Chr.10: 46109495 - 46131678 on Build GRCh38
          Chromosome Location:
          Chr.10: 47501854 - 47523638 on Build GRCh38
          Chromosome Location:
          Chr.10: 46335552 - 46838268 on Build GRCh38
          UniGene ID:
          Hs.314437
          Hs.645213
          Hs.651147
          Hs.447454
          Hs.645346
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s57633

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGAP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276343.2 NP_001263272.2 7 679
          NM_001291379.1 NP_001278308.1 9 871
          NM_133446.3 NP_597703.2 6 610
          XM_011539269.2 XP_011537571.1 4 132
          XM_017015664.1 XP_016871153.1 9 4541
          XM_017015665.1 XP_016871154.1 10 4409
          XM_017015666.1 XP_016871155.1 8 1444
          XM_017015667.1 XP_016871156.1 9 714
          XM_017015668.1 XP_016871157.1 6 330
          XM_017015669.1 XP_016871158.1 6 283
          GenBank mRNA AF411132.1 - 6 610
          Gene Symbol: AGAP12P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_029396.2 7 595
          Gene Symbol: AGAP5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144000.1 NP_001137472.1 7 594
          Gene Symbol: AGAP7P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_126580.1 6 579
          GenBank mRNA AA337655.1 - 2 55
          Gene Symbol: AGAP9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190810.1 NP_001177739.1 7 665
          GenBank mRNA AK160375.1 - 8 3641
          BC171845.1 - 7 665
          Gene Symbol: LOC107984026
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017017029.1 XP_016872518.1 9 1096
          XM_017017030.1 XP_016872519.1 8 1027
          XM_017017031.1 XP_016872520.1 8 1027
          XM_017017032.1 XP_016872521.1 10 1127
          XM_017017033.1 XP_016872522.1 11 890
          XM_017017034.1 XP_016872523.1 10 795
          XM_017017035.1 XP_016872524.1 6 308
          XM_017017036.1 XP_016872525.1 4 469
          XM_017017037.1 XP_016872526.1 6 858
          XM_017017038.1 XP_016872527.1 4 111
          XM_017017039.1 XP_016872528.1 6 327
          XM_017017040.1 XP_016872529.1 6 291

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          GTPase activator activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pw4jl:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0