Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC107984026 SIRNA Assays ›
          Gene Symbol:
          LOC107984026 AGAP4 AGAP6
          Gene Name:
          arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 4
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 6
          Gene Aliases:
          -
          AGAP-4, AGAP-8, AGAP8, CTGLF1, CTGLF5, MRIP2
          AGAP-6, CTGLF3, bA324H6.1
          Chromosome Location:
          Chr.10: 46335552 - 46838268 on Build GRCh38
          Chromosome Location:
          Chr.10: 45825592 - 45858529 on Build GRCh38
          Chromosome Location:
          Chr.10: 49981196 - 50010507 on Build GRCh38
          UniGene ID:
          -
          Hs.314437
          Hs.536158
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s58596

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC107984026
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017017029.1 XP_016872518.1 10 2203
          XM_017017030.1 XP_016872519.1 9 2134
          XM_017017031.1 XP_016872520.1 9 2134
          XM_017017032.1 XP_016872521.1 11 2234
          XM_017017033.1 XP_016872522.1 12 1997
          XM_017017034.1 XP_016872523.1 11 1902
          XM_017017035.1 XP_016872524.1 7 1415
          XM_017017036.1 XP_016872525.1 5 1576
          XM_017017037.1 XP_016872526.1 7 1965
          XM_017017038.1 XP_016872527.1 5 1218
          XM_017017039.1 XP_016872528.1 7 1434
          XM_017017040.1 XP_016872529.1 7 1398
          Gene Symbol: AGAP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276343.2 NP_001263272.2 8 1786
          NM_001291379.1 NP_001278308.1 10 1978
          NM_133446.3 NP_597703.2 7 1717
          XM_011539269.2 XP_011537571.1 6 1324
          XM_017015664.1 XP_016871153.1 10 5648
          XM_017015665.1 XP_016871154.1 11 5516
          XM_017015666.1 XP_016871155.1 9 2551
          XM_017015667.1 XP_016871156.1 10 1821
          XM_017015668.1 XP_016871157.1 8 1571
          XM_017015669.1 XP_016871158.1 8 1479
          GenBank mRNA AF411132.1 - 7 1717
          BC048341.1 - 1 839
          Gene Symbol: AGAP6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077665.2 NP_001071133.2 8 2058
          XM_005269841.3 XP_005269898.1 2 1099
          XM_011539814.2 XP_011538116.1 2 1103
          XM_011539815.2 XP_011538117.1 5 1283
          XM_017016268.1 XP_016871757.1 8 1779
          XM_017016269.1 XP_016871758.1 8 1779
          XM_017016270.1 XP_016871759.1 7 2013
          XM_017016271.1 XP_016871760.1 10 1781
          XM_017016272.1 XP_016871761.1 9 1711
          XM_017016273.1 XP_016871762.1 9 2404
          XM_017016274.1 XP_016871763.1 5 1234

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          GTPase activator activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0