Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATRX SIRNA Assays ›
Gene Symbol:
ATRX
Gene Name:
ATRX, chromatin remodeler
Gene Aliases:
ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX
Chromosome Location:
Chr.X: 77504878 - 77786269 on Build GRCh38
UniGene ID:
Hs.533526
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s59082
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATRX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000489.4
NP_000480.3
9
1059
NM_138270.3
NP_612114.2
8
945
XM_005262153.4
XP_005262210.2
9
1056
XM_005262154.4
XP_005262211.2
9
1059
XM_005262155.4
XP_005262212.2
9
1160
XM_005262156.4
XP_005262213.2
8
912
XM_005262157.4
XP_005262214.2
8
942
XM_006724666.3
XP_006724729.1
8
942
XM_006724667.3
XP_006724730.1
7
798
XM_006724668.2
XP_006724731.1
9
1059
XM_017029601.1
XP_016885090.1
9
1056
XM_017029602.1
XP_016885091.1
9
1140
XM_017029603.1
XP_016885092.1
8
909
XM_017029604.1
XP_016885093.1
8
945
XM_017029605.1
XP_016885094.1
9
1160
XM_017029606.1
XP_016885095.1
8
1046
XM_017029607.1
XP_016885096.1
8
1043
XM_017029608.1
XP_016885097.1
7
795
XM_017029609.1
XP_016885098.1
8
1046
XM_017029610.1
XP_016885099.1
8
1043
XM_017029611.1
XP_016885100.1
7
798
GenBank mRNA
AB102641.1
-
9
791
AB208928.1
-
8
873
AK307791.1
-
8
885
AK308419.1
-
9
998
U72936.2
-
10
1128
U72937.2
-
9
1006
U72938.2
-
8
892
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
DNA methylation
DNA recombination
nucleosome assembly
DNA replication-independent nucleosome assembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
spermatogenesis
positive regulation of nuclear cell cycle DNA replication
covalent chromatin modification
DNA damage response, signal transduction by p53 class mediator
forebrain development
replication fork processing
positive regulation of telomere maintenance
DNA duplex unwinding
post-embryonic forelimb morphogenesis
multicellular organism growth
positive regulation of transcription from RNA polymerase II promoter
Sertoli cell development
protein localization to chromosome, telomeric region
seminiferous tubule development
cellular response to hydroxyurea
regulation of histone H3-K9 trimethylation
negative regulation of telomeric RNA transcription from RNA pol II promoter
positive regulation of telomeric RNA transcription from RNA pol II promoter
negative regulation of maintenance of mitotic sister chromatid cohesion, telomeric
DNA binding
DNA helicase activity
chromatin binding
helicase activity
protein binding
ATP binding
DNA translocase activity
methylated histone binding
histone binding
metal ion binding
chromo shadow domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0