Search
Search

ANKRD36BP2
ANKRD36C| Gene Symbol: ANKRD36BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015424.1 | 14, 15 | 1335 | |||
| GenBank mRNA | BC031698.1 | - | 14, 15 | 1335 | ||
| Gene Symbol: ANKRD36C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310154.1 | NP_001297083.1 | 86, 87 | 6931 | ||