Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36 SIRNA Assays ›
Gene Symbol:
ANKRD36
Gene Name:
ankyrin repeat domain 36
Gene Aliases:
UNQ2430
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
UniGene ID:
Hs.541894
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s61143
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
4
762
XM_006712514.3
XP_006712577.1
4
1072
XM_011511130.1
XP_011509432.1
4
1072
XM_011511131.1
XP_011509433.1
4
1072
XM_017004009.1
XP_016859498.1
4
1072
XM_017004010.1
XP_016859499.1
4
1072
XM_017004011.1
XP_016859500.1
4
1072
XM_017004012.1
XP_016859501.1
4
1072
XM_017004013.1
XP_016859502.1
4
1072
XM_017004014.1
XP_016859503.1
4
1072
XM_017004015.1
XP_016859504.1
4
1072
XM_017004016.1
XP_016859505.1
4
1072
XM_017004017.1
XP_016859506.1
4
1072
XM_017004018.1
XP_016859507.1
4
1072
XM_017004019.1
XP_016859508.1
4
1072
XM_017004020.1
XP_016859509.1
4
1072
XM_017004021.1
XP_016859510.1
4
1072
XM_017004022.1
XP_016859511.1
4
1072
XM_017004023.1
XP_016859512.1
4
1072
XM_017004024.1
XP_016859513.1
4
1072
XM_017004025.1
XP_016859514.1
4
1072
XM_017004026.1
XP_016859515.1
4
1072
XM_017004027.1
XP_016859516.1
4
1072
XM_017004028.1
XP_016859517.1
4
1072
XM_017004029.1
XP_016859518.1
4
1072
XM_017004030.1
XP_016859519.1
4
1072
XM_017004031.1
XP_016859520.1
4
1072
XM_017004032.1
XP_016859521.1
4
1072
XM_017004033.1
XP_016859522.1
4
1072
XM_017004034.1
XP_016859523.1
4
1072
XM_017004035.1
XP_016859524.1
4
1072
XM_017004036.1
XP_016859525.1
4
1072
XM_017004037.1
XP_016859526.1
4
1072
XM_017004038.1
XP_016859527.1
4
1072
XM_017004039.1
XP_016859528.1
4
1072
XM_017004040.1
XP_016859529.1
4
1072
XM_017004041.1
XP_016859530.1
4
1072
XM_017004042.1
XP_016859531.1
4
1072
XM_017004043.1
XP_016859532.1
4
1072
XM_017004044.1
XP_016859533.1
4
1072
XM_017004045.1
XP_016859534.1
4
1072
XM_017004049.1
XP_016859538.1
4
1072
GenBank mRNA
BC128262.1
-
4
601
BC128517.1
-
4
601
40118468
BX647769.1
-
4
707
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0