Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015696.4 | NP_056511.2 | 1 | 157 | ||
GenBank mRNA | AF320068.2 | - | 1 | 156 | ||
AK027683.1 | - | 1 | 157 | |||
AY358402.1 | - | 1 | 151 | |||
BC032788.1 | - | 1 | 130 | 5212085 | ||
BI550170.1 | - | 1 | 162 |