Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GRB2 SIRNA Assays ›
          Gene Symbol:
          GRB2
          Gene Name:
          growth factor receptor bound protein 2
          Gene Aliases:
          ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084, NCKAP2
          Chromosome Location:
          Chr.17: 75318076 - 75405709 on Build GRCh38
          UniGene ID:
          Hs.444356
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s6119

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GRB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002086.4 NP_002077.1 3 514
          NM_203506.2 NP_987102.1 3 515
          GenBank mRNA AF498925.1 - 2 157
          AK091010.1 - 3 942
          AK315565.1 - 3 510
          BC000631.2 - 3 459 3345524
          BI459414.1 - 3 521
          CR450363.1 - 2 157
          CR541942.1 - 2 157
          DA458680.1 - 3 515
          L29511.1 - 2 193
          M96995.1 - 2 235

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          epidermal growth factor receptor signaling pathway
          Ras protein signal transduction
          cell-cell signaling
          axon guidance
          insulin receptor signaling pathway
          fibroblast growth factor receptor signaling pathway
          positive regulation of signal transduction
          regulation of phosphatidylinositol 3-kinase signaling
          viral process
          positive regulation of actin filament polymerization
          T cell costimulation
          receptor internalization
          phosphatidylinositol-3-phosphate biosynthetic process
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          ERBB2 signaling pathway
          negative regulation of epidermal growth factor receptor signaling pathway
          signal transduction in response to DNA damage
          regulation of MAPK cascade
          positive regulation of GTPase activity
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          anatomical structure formation involved in morphogenesis
          leukocyte migration
          branching involved in labyrinthine layer morphogenesis
          cellular response to ionizing radiation
          positive regulation of reactive oxygen species metabolic process
          SH3/SH2 adaptor activity
          Ras guanyl-nucleotide exchange factor activity
          epidermal growth factor receptor binding
          neurotrophin TRKA receptor binding
          protein binding
          1-phosphatidylinositol-3-kinase activity
          SH3 domain binding
          protein kinase binding
          protein phosphatase binding
          identical protein binding
          insulin receptor substrate binding
          poly(A) RNA binding
          ephrin receptor binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0