Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABL2 SIRNA Assays ›
          Gene Symbol:
          Abl2
          Gene Name:
          v-abl Abelson murine leukemia viral oncogene 2 (arg, Abelson-related gene)
          Gene Aliases:
          AA536808, Abll, Arg
          Chromosome Location:
          Chr.1: 156558171 - 156649619 on Build GRCm38
          UniGene ID:
          Mm.329515
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s61807

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Abl2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136104.1 NP_001129576.1 9 1698
          NM_009595.3 NP_033725.2 9 1698
          GenBank mRNA BC137771.1 - 9 1498 8860790
          BC144896.1 - 9 1498
          BC144897.1 - 9 1498 9053445
          DQ084361.1 - 9 1436
          U40827.1 - 5 513

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          phagocytosis
          substrate-dependent cell migration, cell extension
          actin filament organization
          cell adhesion
          epidermal growth factor receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          learning
          visual learning
          positive regulation of phospholipase C activity
          positive regulation of neuron projection development
          phosphorylation
          neuron remodeling
          cell migration
          peptidyl-tyrosine phosphorylation
          cerebellum morphogenesis
          negative regulation of cell-cell adhesion
          actin cytoskeleton organization
          positive regulation of protein binding
          cellular protein localization
          negative regulation of Rho protein signal transduction
          exploration behavior
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          alpha-beta T cell differentiation
          platelet-derived growth factor receptor signaling pathway
          dendrite morphogenesis
          neuromuscular process controlling balance
          actin filament bundle assembly
          positive regulation of oxidoreductase activity
          Bergmann glial cell differentiation
          positive regulation of ERK1 and ERK2 cascade
          cellular response to retinoic acid
          dendritic spine maintenance
          positive regulation of interleukin-2 secretion
          positive regulation of interferon-gamma secretion
          regulation of extracellular matrix organization
          positive regulation of Wnt signaling pathway, planar cell polarity pathway
          negative regulation of endothelial cell apoptotic process
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          manganese ion binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0