Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AGT SIRNA Assays ›
Gene Symbol:
Agt
Gene Name:
angiotensinogen (serpin peptidase inhibitor, clade A, member 8)
Gene Aliases:
AI265500, AngI, AngII, Aogen, Serpina8
Chromosome Location:
Chr.8: 124556587 - 124569707 on Build GRCm38
UniGene ID:
Mm.301626
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s62141
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Agt
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007428.3
NP_031454.3
3
1092
GenBank mRNA
AK005354.1
-
3
1090
AK018763.1
-
3
1090
AK139179.1
-
3
1091
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle rupture
blood vessel development
branching involved in ureteric bud morphogenesis
kidney development
regulation of systemic arterial blood pressure by circulatory renin-angiotensin
angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure
renal response to blood flow involved in circulatory renin-angiotensin regulation of systemic arterial blood pressure
renin-angiotensin regulation of aldosterone production
regulation of renal output by angiotensin
regulation of heart rate
brain renin-angiotensin system
renal system process
angiotensin-mediated drinking behavior
positive regulation of extracellular matrix constituent secretion
cellular sodium ion homeostasis
cell-matrix adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
activation of phospholipase C activity
activation of NF-kappaB-inducing kinase activity
excretion
establishment of blood-nerve barrier
regulation of blood pressure
positive regulation of cell proliferation
negative regulation of cell proliferation
response to cold
response to salt stress
regulation of gene expression
positive regulation of activation of JAK2 kinase activity
positive regulation of endothelial cell migration
positive regulation of cardiac muscle hypertrophy
positive regulation of gene expression
positive regulation of cholesterol esterification
negative regulation of endopeptidase activity
regulation of norepinephrine secretion
positive regulation of phosphatidylinositol 3-kinase signaling
artery smooth muscle contraction
response to muscle activity involved in regulation of muscle adaptation
negative regulation of angiogenesis
extracellular matrix organization
negative regulation of cell growth
peristalsis
positive regulation of cellular protein metabolic process
positive regulation of superoxide anion generation
positive regulation of peptidyl-serine phosphorylation
negative regulation of tissue remodeling
catenin import into nucleus
regulation of renal sodium excretion
positive regulation of renal sodium excretion
positive regulation of multicellular organism growth
vasoconstriction
vasodilation
hormone metabolic process
drinking behavior
regulation of apoptotic process
positive regulation of catalytic activity
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
positive regulation of nitric oxide biosynthetic process
positive regulation of fatty acid biosynthetic process
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of blood pressure
positive regulation of transcription, DNA-templated
positive regulation of vasodilation
positive regulation of organ growth
astrocyte activation
fibroblast proliferation
positive regulation of fibroblast proliferation
regulation of long-term neuronal synaptic plasticity
smooth muscle cell proliferation
cytokine secretion
regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
smooth muscle cell differentiation
negative regulation of neurotrophin TRK receptor signaling pathway
stress-activated MAPK cascade
regulation of calcium ion transport
regulation of transmission of nerve impulse
cell growth involved in cardiac muscle cell development
positive regulation of protein tyrosine kinase activity
ERK1 and ERK2 cascade
uterine smooth muscle contraction
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of protein kinase C activity
regulation of extracellular matrix assembly
positive regulation of membrane hyperpolarization
positive regulation of gap junction assembly
regulation of cardiac conduction
positive regulation of extrinsic apoptotic signaling pathway
serine-type endopeptidase inhibitor activity
hormone activity
sodium channel regulator activity
angiotensin receptor binding
type 1 angiotensin receptor binding
type 2 angiotensin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline