Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AGTR1A SIRNA Assays ›
Gene Symbol:
Agtr1a
Gene Name:
angiotensin II receptor, type 1a
Gene Aliases:
1810074K20Rik, AG2S, AI551199, AT1, AT1a, AT2R1, AT2R1A, Agtr-1a, Agtr1, Angtr-1a
Chromosome Location:
Chr.13: 30336356 - 30382867 on Build GRCm38
UniGene ID:
Mm.35062
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s62145
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Agtr1a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_177322.3
NP_796296.1
3
598
GenBank mRNA
BC036175.1
-
3
497
4989471
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_030985, XM_008771594, XM_006253882, XM_008771593, BC078810, and others.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
kidney development
positive regulation of receptor recycling
regulation of systemic arterial blood pressure by circulatory renin-angiotensin
renin secretion into blood stream
renin-angiotensin regulation of aldosterone production
regulation of renal output by angiotensin
brain renin-angiotensin system
regulation of pH
inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
Rho protein signal transduction
heart development
positive regulation of cell proliferation
positive regulation of cholesterol esterification
regulation of vasoconstriction
calcium-mediated signaling
positive regulation of cellular protein metabolic process
positive regulation of phospholipase A2 activity
positive regulation of superoxide anion generation
regulation of smooth muscle cell apoptotic process
negative regulation of smooth muscle cell apoptotic process
angiotensin-activated signaling pathway
vasoconstriction
dopamine biosynthetic process
drinking behavior
activation of Janus kinase activity
negative regulation of neuron apoptotic process
response to estrogen
positive regulation of blood pressure
positive regulation of cytokine secretion
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
cell chemotaxis
cellular response to dexamethasone stimulus
phospholipase C-activating angiotensin-activated signaling pathway
positive regulation of branching involved in ureteric bud morphogenesis
angiotensin type I receptor activity
G-protein alpha-subunit binding
signal transducer activity
G-protein coupled receptor activity
angiotensin type II receptor activity
protein binding
peptide hormone binding
protein kinase binding
bradykinin receptor binding
D1 dopamine receptor binding
protein self-association
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wfn85:80/100.66.78.97:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0