Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AHR SIRNA Assays ›
          Gene Symbol:
          Ahr
          Gene Name:
          aryl-hydrocarbon receptor
          Gene Aliases:
          Ah, Ahh, Ahre, In, bHLHe76
          Chromosome Location:
          Chr.12: 35497979 - 35534989 on Build GRCm38
          UniGene ID:
          Mm.341377
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s62161

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ahr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013464.4 NP_038492.1 10 1533
          GenBank mRNA AF405560.1 - 10 1167
          AF405561.1 - 10 1167
          AF405562.1 - 10 1167
          AF405563.1 - 10 1167
          AF405565.1 - 10 1167
          AF405566.1 - 10 1167
          AF405567.1 - 10 1167
          AF405568.1 - 10 1167
          AF405570.1 - 10 1167
          AK136072.1 - 4 417
          AK153678.1 - 10 1534
          AK155380.1 - 10 1585
          AY662080.1 - 10 1167
          AY662081.1 - 9 1063
          AY662082.1 - 10 1167
          AY662083.1 - 10 1167
          AY662084.1 - 10 1167
          D38416.1 - 10 1182

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          ovarian follicle development
          blood vessel development
          patterning of blood vessels
          B cell homeostasis
          liver development
          B-1 B cell homeostasis
          blood vessel remodeling
          lymphocyte homeostasis
          immune system process
          negative regulation of systemic arterial blood pressure
          cardiac left ventricle morphogenesis
          circumferential growth involved in left ventricle morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          xenobiotic metabolic process
          response to stress
          cell cycle
          signal transduction
          blood circulation
          regulation of blood pressure
          response to xenobiotic stimulus
          response to toxic substance
          regulation of gene expression
          response to organic cyclic compound
          B cell differentiation
          intracellular receptor signaling pathway
          prostate gland development
          regulation of B cell proliferation
          circadian regulation of gene expression
          embryonic hemopoiesis
          post-embryonic hemopoiesis
          camera-type eye development
          T cell homeostasis
          positive regulation of cell size
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
          negative regulation of vasoconstriction
          positive regulation of growth
          rhythmic process
          blood vessel morphogenesis
          spleen development
          reproductive structure development
          gland development
          smooth muscle tissue development
          regulation of blood vessel size
          regulation of heart growth
          negative regulation of necrotic cell death
          venous blood vessel development
          kidney morphogenesis
          common bile duct development
          cellular response to cAMP
          glomerulus morphogenesis
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          signal transducer activity
          aryl hydrocarbon receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          transcription factor binding
          enhancer binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          protein dimerization activity
          Hsp90 protein binding
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0