Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GSN SIRNA Assays ›
Gene Symbol:
GSN
Gene Name:
gelsolin
Gene Aliases:
ADF, AGEL
Chromosome Location:
Chr.9: 121201082 - 121332844 on Build GRCh38
UniGene ID:
Hs.522373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6242
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GSN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000177.4
NP_000168.1
17
2358
NM_001127662.1
NP_001121134.1
17
2271
NM_001127663.1
NP_001121135.2
19
2476
NM_001127664.1
NP_001121136.1
19
2443
NM_001127665.1
NP_001121137.1
19
2438
NM_001127666.1
NP_001121138.1
19
2435
NM_001127667.1
NP_001121139.1
19
2426
NM_001258029.1
NP_001244958.1
17
2258
NM_001258030.1
NP_001244959.1
17
2335
NM_198252.2
NP_937895.1
18
2364
XM_005251943.1
XP_005252000.1
18
2462
XM_005251944.1
XP_005252001.1
18
2453
XM_005251945.3
XP_005252002.1
18
2366
XM_006717079.1
XP_006717142.1
18
2300
XM_011518584.1
XP_011516886.1
19
2453
XM_011518585.1
XP_011516887.1
17
2273
XM_011518586.1
XP_011516888.1
20
2476
XM_011518587.2
XP_011516889.1
20
2518
XM_011518588.2
XP_011516890.1
20
2480
XM_011518589.2
XP_011516891.1
20
2476
XM_011518590.2
XP_011516892.1
20
2428
XM_011518591.2
XP_011516893.1
25
3520
XM_011518592.1
XP_011516894.1
19
2342
XM_011518593.1
XP_011516895.1
20
2488
XM_011518594.1
XP_011516896.1
14
1685
XM_017014643.1
XP_016870132.1
26
3671
XM_017014644.1
XP_016870133.1
26
3605
XM_017014645.1
XP_016870134.1
18
2417
XM_017014646.1
XP_016870135.1
21
2588
XM_017014647.1
XP_016870136.1
27
3804
XM_017014648.1
XP_016870137.1
20
2415
GenBank mRNA
AK096280.1
-
18
2337
AK125819.1
-
26
3167
AK295201.1
-
18
2202
AK295572.1
-
19
2397
AK297870.1
-
17
2216
AK299453.1
-
17
2323
AK304629.1
-
17
2279
AK304675.1
-
16
2312
AK307859.1
-
17
2221
AK315494.1
-
17
2329
BC017491.1
-
7
808
BC026033.1
-
17
2302
4661084
BX647999.1
-
17
2260
X04412.1
-
17
2311
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis, engulfment
aging
positive regulation of gene expression
oligodendrocyte development
striated muscle atrophy
extracellular matrix disassembly
actin filament polymerization
protein destabilization
tissue regeneration
sequestering of actin monomers
cellular protein metabolic process
actin nucleation
response to ethanol
negative regulation of viral entry into host cell
phosphatidylinositol-mediated signaling
actin filament severing
barbed-end actin filament capping
positive regulation of actin nucleation
response to folic acid
actin filament capping
cilium morphogenesis
cellular response to cadmium ion
regulation of podosome assembly
actin filament reorganization
renal protein absorption
hepatocyte apoptotic process
positive regulation of keratinocyte apoptotic process
regulation of wound healing, spreading of epidermal cells
regulation of establishment of T cell polarity
regulation of plasma membrane raft polarization
regulation of receptor clustering
positive regulation of protein processing in phagocytic vesicle
amyloid fibril formation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
actin binding
calcium ion binding
protein binding
protein domain specific binding
myosin II binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0