Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AQP1 SIRNA Assays ›
          Gene Symbol:
          Aqp1
          Gene Name:
          aquaporin 1
          Gene Aliases:
          CHIP28
          Chromosome Location:
          Chr.6: 55336299 - 55348555 on Build GRCm38
          UniGene ID:
          Mm.18625
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s62520

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Aqp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007472.2 NP_031498.1 1 281
          GenBank mRNA AK081886.1 - 1 149
          AK086688.1 - 1 149
          AK128905.1 - 1 148
          AK157333.1 - 1 149
          AK158226.1 - 1 149
          AK158389.1 - 1 149
          AK171627.1 - 1 149
          AK172361.1 - 1 149
          BB850287.1 - 1 282
          L02914.1 - 1 148

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          glomerular filtration
          renal water transport
          cGMP biosynthetic process
          transport
          potassium ion transport
          water transport
          cell volume homeostasis
          cellular water homeostasis
          positive regulation of lamellipodium assembly
          positive regulation of epithelial cell migration
          carbon dioxide transport
          organic cation transport
          ammonium transport
          glycerol transport
          sensory perception of pain
          cellular homeostasis
          water homeostasis
          nitric oxide transport
          positive regulation of cell migration
          establishment or maintenance of actin cytoskeleton polarity
          secretion by cell
          secretory granule organization
          cellular response to stress
          carbohydrate transmembrane transport
          cellular response to UV
          transepithelial water transport
          carbon dioxide transmembrane transport
          wound healing
          response to drug
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          lipid digestion
          positive regulation of angiogenesis
          positive regulation of saliva secretion
          positive regulation of fibroblast proliferation
          camera-type eye morphogenesis
          corticotropin secretion
          transmembrane transport
          renal water absorption
          cellular response to hydrogen peroxide
          cellular response to inorganic substance
          cellular response to mechanical stimulus
          cellular response to copper ion
          cellular response to mercury ion
          cellular response to retinoic acid
          cellular response to cAMP
          cellular response to hypoxia
          cellular response to salt stress
          cellular hyperosmotic response
          cellular response to dexamethasone stimulus
          cellular response to nitric oxide
          potassium ion transmembrane transport
          ammonium transmembrane transport
          maintenance of symbiont-containing vacuole by host
          cation transmembrane transport
          transporter activity
          intracellular cGMP activated cation channel activity
          potassium channel activity
          water transmembrane transporter activity
          ammonium transmembrane transporter activity
          potassium ion transmembrane transporter activity
          glycerol transmembrane transporter activity
          water channel activity
          glycerol channel activity
          transmembrane transporter activity
          nitric oxide transmembrane transporter activity
          carbon dioxide transmembrane transporter activity
          ephrin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0