Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTNNB1 SIRNA Assays ›
          Gene Symbol:
          Ctnnb1
          Gene Name:
          catenin (cadherin associated protein), beta 1
          Gene Aliases:
          Bfc, Catnb, Mesc
          Chromosome Location:
          Chr.9: 120933400 - 120960507 on Build GRCm38
          UniGene ID:
          Mm.291928
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s63417

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ctnnb1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001165902.1 NP_001159374.1 9 1376
          NM_007614.3 NP_031640.1 9 1576
          GenBank mRNA AK018515.1 - 9 1490
          AK020013.1 - 9 1492
          AK035311.1 - 9 1489
          AK132398.1 - 9 1557
          AK133656.1 - 9 1489
          AK135743.1 - 6 1043
          AK184861.1 - 3 226
          AY751549.1 - 9 1501
          BC043078.1 - 6 978
          BC043481.1 - 5 610
          BC048153.1 - 9 1501 5709247
          BC053065.1 - 9 1501 5709247
          M90364.1 - 9 1386

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053357, XM_008766691, AF121265, FQ224468, FQ222424.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          embryonic axis specification
          cell morphogenesis involved in differentiation
          skeletal system development
          patterning of blood vessels
          vasculogenesis
          branching involved in ureteric bud morphogenesis
          in utero embryonic development
          gastrulation with mouth forming second
          endoderm formation
          cell fate specification
          cell fate determination
          endodermal cell fate commitment
          neuron migration
          kidney development
          neural plate development
          vasculature development
          positive regulation of neuroblast proliferation
          positive regulation of mesenchymal cell proliferation
          lens morphogenesis in camera-type eye
          ventricular compact myocardium morphogenesis
          regulation of secondary heart field cardioblast proliferation
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          metanephros morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          cell adhesion
          cell-matrix adhesion
          chemical synaptic transmission
          ectoderm development
          nervous system development
          glial cell fate determination
          heart development
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          anterior/posterior axis specification
          dorsal/ventral axis specification
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          cellular process
          regulation of gene expression
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of heparan sulfate proteoglycan biosynthetic process
          Schwann cell proliferation
          Wnt signaling pathway
          morphogenesis of embryonic epithelium
          single organismal cell-cell adhesion
          stem cell population maintenance
          layer formation in cerebral cortex
          central nervous system vasculogenesis
          hair cycle process
          hemopoiesis
          cell differentiation
          T cell differentiation
          osteoclast differentiation
          lung development
          male genitalia development
          regulation of epithelial cell differentiation
          positive regulation of epithelial cell differentiation
          forebrain development
          regulation of centriole-centriole cohesion
          pancreas development
          hair follicle morphogenesis
          regulation of myelination
          positive regulation of telomere maintenance via telomerase
          negative regulation of chondrocyte differentiation
          response to estradiol
          T cell differentiation in thymus
          negative regulation of protein sumoylation
          response to cytokine
          adherens junction organization
          adherens junction assembly
          protein localization to cell surface
          cellular protein localization
          embryonic heart tube development
          genitalia morphogenesis
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          embryonic skeletal limb joint morphogenesis
          regulation of cell proliferation
          regulation of T cell proliferation
          odontogenesis of dentin-containing tooth
          regulation of cell fate specification
          embryonic digit morphogenesis
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of MAPK cascade
          skin development
          response to estrogen
          canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          bone resorption
          regulation of cell differentiation
          negative regulation of cell differentiation
          positive regulation of endothelial cell differentiation
          regulation of osteoblast differentiation
          positive regulation of osteoblast differentiation
          regulation of osteoclast differentiation
          negative regulation of osteoclast differentiation
          positive regulation of fibroblast growth factor receptor signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of mitotic cell cycle, embryonic
          chromatin-mediated maintenance of transcription
          cell maturation
          synaptic vesicle transport
          animal organ development
          thymus development
          oocyte development
          embryonic foregut morphogenesis
          regulation of smooth muscle cell proliferation
          negative regulation of oligodendrocyte differentiation
          synapse organization
          smooth muscle cell differentiation
          protein heterooligomerization
          positive regulation of telomerase activity
          cardiac muscle cell proliferation
          oviduct development
          canonical Wnt signaling pathway
          limb development
          trachea morphogenesis
          trachea formation
          epithelial tube branching involved in lung morphogenesis
          lung cell differentiation
          lung-associated mesenchyme development
          mesenchyme development
          lung induction
          epithelial cell differentiation involved in prostate gland development
          positive regulation of epithelial cell proliferation involved in prostate gland development
          hair follicle placode formation
          mesenchymal cell proliferation involved in lung development
          cardiac vascular smooth muscle cell differentiation
          coronary artery morphogenesis
          epicardium-derived cardiac vascular smooth muscle cell differentiation
          positive regulation of branching involved in lung morphogenesis
          endothelial tube morphogenesis
          fungiform papilla formation
          canonical Wnt signaling pathway involved in positive regulation of cardiac outflow tract cell proliferation
          sympathetic ganglion development
          regulation of centromeric sister chromatid cohesion
          cellular response to mechanical stimulus
          cellular response to growth factor stimulus
          cellular response to organic cyclic compound
          cellular response to indole-3-methanol
          renal system development
          renal vesicle formation
          renal inner medulla development
          renal outer medulla development
          nephron tubule formation
          mesenchyme morphogenesis
          regulation of nephron tubule epithelial cell differentiation
          regulation of calcium ion import
          negative regulation of neuron death
          cellular response to insulin-like growth factor stimulus
          regulation of protein localization to cell surface
          positive regulation of determination of dorsal identity
          negative regulation of apoptotic signaling pathway
          RNA polymerase II activating transcription factor binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          signal transducer activity
          protein binding
          protein C-terminus binding
          transcription factor binding
          enzyme binding
          kinase binding
          protein phosphatase binding
          estrogen receptor binding
          protein complex binding
          ionotropic glutamate receptor binding
          nuclear hormone receptor binding
          transcription regulatory region DNA binding
          ion channel binding
          alpha-catenin binding
          cadherin binding
          SMAD binding
          nitric-oxide synthase binding
          I-SMAD binding
          R-SMAD binding
          repressing transcription factor binding
          euchromatin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0