Search
Search

Csf2rb| Gene Symbol: Csf2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007780.4 | NP_031806.3 | 4 | 586 | ||
| GenBank mRNA | AF483496.1 | - | 3 | 328 | ||
| AF483497.1 | - | 3 | 328 | |||
| AK133420.1 | - | 530 | ||||
| AK152608.1 | - | 4 | 526 | |||
| AK154286.1 | - | 4 | 523 | |||
| AK155109.1 | - | 4 | 557 | |||
| AK155215.1 | - | 4 | 520 | |||
| AK155699.1 | - | 4 | 523 | |||
| AK155803.1 | - | 4 | 523 | |||
| AK155822.1 | - | 4 | 526 | |||
| AK155885.1 | - | 4 | 527 | |||
| AK171477.1 | - | 4 | 523 | |||
| BC113202.1 | - | 5 | 597 | 40048156 | ||
| M34397.1 | - | 5 | 615 | |||