Search
Search

Dscr3| Gene Symbol: Dscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007834.3 | NP_031860.1 | 7 | 932 | ||
| GenBank mRNA | AB001990.1 | - | 7 | 747 | ||
| AK077485.1 | - | 7 | 746 | |||
| AK077628.1 | - | 7 | 738 | |||
| AK219359.1 | - | 1 | 56 | |||
| BC003740.1 | - | 7 | 720 | 3592608 | ||