Search
Search

Enpep| Gene Symbol: Enpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007934.3 | NP_031960.1 | 1 | 724 | ||
| GenBank mRNA | AK034613.1 | - | 1 | 696 | ||
| AK128975.1 | - | 1 | 681 | |||
| AK146808.1 | - | 1 | 696 | |||
| BC127060.1 | - | 1 | 588 | 40109863 | ||
| DQ000292.1 | - | 1 | 464 | |||
| M29961.1 | - | 1 | 543 | |||