Search
Search

Fgr| Gene Symbol: Fgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010208.4 | NP_034338.3 | 9, 10 | 1224 | ||
| GenBank mRNA | AK036438.1 | - | 9, 10 | 1146 | ||
| AK036476.1 | - | 9, 10 | 1396 | |||
| AK038141.1 | - | 9, 10 | 1130 | |||
| AK089812.1 | - | 9, 10 | 1194 | |||
| AK155902.1 | - | 9, 10 | 1200 | |||
| AK157640.1 | - | 1178 | ||||
| AK171824.1 | - | 9, 10 | 1178 | |||
| BC057863.1 | - | 9, 10 | 1093 | 5317449 | ||
| X16440.1 | - | 9, 10 | 1159 | |||
| X52191.1 | - | 9, 10 | 1131 | |||