Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HMGB1 SIRNA Assays ›
          Gene Symbol:
          HMGB1
          Gene Name:
          high mobility group box 1
          Gene Aliases:
          HMG-1, HMG1, HMG3, SBP-1
          Chromosome Location:
          Chr.13: 30457916 - 30617597 on Build GRCh38
          UniGene ID:
          Hs.434102
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s6647

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HMGB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001313892.1 NP_001300821.1 5 1477
          NM_001313893.1 NP_001300822.1 5 2237
          NM_002128.5 NP_002119.1 5 1480
          GenBank mRNA AK057120.1 - 5 1342
          AK122825.1 - 5 2238
          AK291494.1 - 5 1449
          AL110194.1 - 1 77
          BC030981.1 - 5 1390 4041682
          BC141844.1 - 5 1371 8860319
          CR749614.1 - 5 1468

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          eye development
          myeloid dendritic cell activation
          endothelial cell proliferation
          activation of innate immune response
          plasmacytoid dendritic cell activation
          macrophage activation involved in immune response
          dendritic cell chemotaxis
          inflammatory response to antigenic stimulus
          regulation of tolerance induction
          regulation of T cell mediated immune response to tumor cell
          DNA topological change
          base-excision repair
          apoptotic DNA fragmentation
          DNA recombination
          regulation of transcription from RNA polymerase II promoter
          autophagy
          inflammatory response
          positive regulation of cytosolic calcium ion concentration
          regulation of autophagy
          negative regulation of RNA polymerase II transcriptional preinitiation complex assembly
          lung development
          neuron projection development
          chromatin assembly
          regulation of restriction endodeoxyribonuclease activity
          activation of protein kinase activity
          DNA geometric change
          positive regulation of mismatch repair
          negative regulation of interferon-gamma production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of tumor necrosis factor production
          V(D)J recombination
          positive regulation of toll-like receptor 2 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          positive regulation of toll-like receptor 9 signaling pathway
          T-helper 1 cell activation
          endothelial cell chemotaxis
          positive regulation of activated T cell proliferation
          positive regulation of apoptotic process
          apoptotic cell clearance
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of CD4-positive, alpha-beta T cell differentiation
          positive regulation of DNA binding
          positive regulation of MAPK cascade
          negative regulation of blood vessel endothelial cell migration
          T-helper 1 cell differentiation
          innate immune response
          positive regulation of myeloid cell differentiation
          positive regulation of glycogen catabolic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of interleukin-1 secretion
          positive regulation of interleukin-1 beta secretion
          positive chemotaxis
          DNA ligation involved in DNA repair
          positive regulation of DNA ligation
          response to glucocorticoid
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of monocyte chemotaxis
          positive regulation of wound healing
          neutrophil clearance
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of sprouting angiogenesis
          tumor necrosis factor secretion
          negative regulation of apoptotic cell clearance
          positive regulation of interleukin-6 secretion
          regulation of nucleotide-excision repair
          positive regulation of dendritic cell differentiation
          four-way junction DNA binding
          bubble DNA binding
          lipopolysaccharide binding
          phosphatidylserine binding
          damaged DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          cytokine activity
          protein binding
          transcription factor binding
          DNA binding, bending
          calcium-dependent protein kinase regulator activity
          lyase activity
          C-X-C chemokine binding
          protein kinase activator activity
          chemoattractant activity
          poly(A) RNA binding
          RAGE receptor binding
          DNA polymerase binding
          repressing transcription factor binding
          supercoiled DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0