Search
Search

Glul| Gene Symbol: Glul | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008131.4 | NP_032157.2 | 3 | 373 | ||
| GenBank mRNA | AK137969.1 | - | 3 | 360 | ||
| AK159106.1 | - | 3 | 359 | |||
| AK160670.1 | - | 3 | 360 | |||
| AK162685.1 | - | 3 | 358 | |||
| AK168493.1 | - | 3 | 357 | |||
| AY044241.1 | - | 3 | 357 | |||
| BC015086.1 | - | 3 | 357 | 4224865 | ||
| U09114.1 | - | 2 | 249 | |||
| X16314.1 | - | 3 | 334 | |||