Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GRIN1 SIRNA Assays ›
          Gene Symbol:
          Grin1
          Gene Name:
          glutamate receptor, ionotropic, NMDA1 (zeta 1)
          Gene Aliases:
          GluN1, GluRdelta1, GluRzeta1, M100174, NMD-R1, NMDAR1, NR1, Nmdar, Rgsc174
          Chromosome Location:
          Chr.2: 25291177 - 25319198 on Build GRCm38
          UniGene ID:
          Mm.278672
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s67051

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Grin1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177656.2 NP_001171127.1 13 2015
          NM_001177657.2 NP_001171128.1 12 1952
          NM_008169.3 NP_032195.1 12 1952
          GenBank mRNA AK158140.1 - 12 1892
          AK158175.1 - 13 1953
          BC039157.1 - 12 1868 4507986
          D10028.1 - 12 1744

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          conditioned taste aversion
          startle response
          suckling behavior
          response to amphetamine
          transport
          ion transport
          cation transport
          calcium ion transport
          cellular calcium ion homeostasis
          chemical synaptic transmission
          respiratory gaseous exchange
          learning or memory
          learning
          memory
          long-term memory
          associative learning
          adult locomotory behavior
          olfactory learning
          visual learning
          regulation of cell communication
          positive regulation of cell death
          propylene metabolic process
          sensory perception of pain
          pons maturation
          cerebral cortex development
          ion transmembrane transport
          regulation of ion transmembrane transport
          social behavior
          ionotropic glutamate receptor signaling pathway
          synaptic transmission, glutamatergic
          regulation of membrane potential
          positive regulation of apoptotic process
          response to morphine
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          regulation of respiratory gaseous exchange
          response to ethanol
          positive regulation of transcription from RNA polymerase II promoter
          regulation of synaptic plasticity
          regulation of neuronal synaptic plasticity
          regulation of long-term neuronal synaptic plasticity
          rhythmic process
          regulation of dendrite morphogenesis
          regulation of axonogenesis
          neuromuscular process
          protein tetramerization
          regulation of synapse assembly
          calcium ion homeostasis
          excitatory postsynaptic potential
          prepulse inhibition
          male mating behavior
          calcium ion transmembrane transport
          cation transmembrane transport
          positive regulation of excitatory postsynaptic potential
          receptor activity
          ionotropic glutamate receptor activity
          NMDA glutamate receptor activity
          receptor binding
          ion channel activity
          extracellular-glutamate-gated ion channel activity
          cation channel activity
          calcium channel activity
          calcium ion binding
          protein binding
          calmodulin binding
          glycine binding
          glutamate binding
          enzyme binding
          voltage-gated cation channel activity
          glutamate receptor binding
          neurotransmitter binding
          protein heterodimerization activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0