Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HES1 SIRNA Assays ›
Gene Symbol:
Hes1
Gene Name:
hairy and enhancer of split 1 (Drosophila)
Gene Aliases:
Hry, bHLHb39
Chromosome Location:
Chr.16: 30065357 - 30067796 on Build GRCm38
UniGene ID:
Mm.390859
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s67463
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hes1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008235.2
NP_032261.1
1
310
GenBank mRNA
AK133538.1
-
1
326
BC018375.1
-
1
310
4505427
BC051428.1
-
1
311
6478994
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_024360, D13417, BC061730, L04527.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
liver development
embryonic heart tube morphogenesis
outflow tract morphogenesis
regulation of secondary heart field cardioblast proliferation
ventricular septum development
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein complex assembly
cell adhesion
Notch signaling pathway
smoothened signaling pathway
STAT protein import into nucleus
pattern specification process
positive regulation of cell proliferation
positive regulation of gene expression
negative regulation of gene expression
negative regulation of neuron projection development
cell migration
telencephalon development
midbrain-hindbrain boundary morphogenesis
oculomotor nerve development
trochlear nerve development
hindbrain morphogenesis
forebrain radial glial cell differentiation
neural tube development
pituitary gland development
adenohypophysis development
lung development
positive regulation of BMP signaling pathway
midbrain development
pancreas development
somatic stem cell population maintenance
hair cell differentiation
aorta morphogenesis
ascending aorta morphogenesis
positive regulation of T cell proliferation
auditory receptor cell differentiation
positive regulation of tyrosine phosphorylation of Stat3 protein
auditory receptor cell fate determination
positive regulation of DNA binding
regulation of protein heterodimerization activity
cell fate commitment
negative regulation of cell differentiation
regulation of fat cell differentiation
negative regulation of auditory receptor cell differentiation
negative regulation of neuron differentiation
positive regulation of glial cell differentiation
positive regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of mitotic cell cycle, embryonic
lateral inhibition
positive regulation of JAK-STAT cascade
cell maturation
regulation of timing of cell differentiation
thymus development
cell morphogenesis involved in neuron differentiation
positive regulation of astrocyte differentiation
negative regulation of oligodendrocyte differentiation
artery morphogenesis
regulation of neurogenesis
negative regulation of neurogenesis
regulation of timing of neuron differentiation
negative regulation of glial cell proliferation
ventricular septum morphogenesis
labyrinthine layer blood vessel development
common bile duct development
negative regulation of stomach neuroendocrine cell differentiation
cardiac neural crest cell development involved in outflow tract morphogenesis
pharyngeal arch artery morphogenesis
cochlea development
negative regulation of calcium ion import
vascular smooth muscle cell development
neuronal stem cell population maintenance
negative regulation of RNA polymerase II regulatory region sequence-specific DNA binding
positive regulation of protein targeting to mitochondrion
cellular response to nerve growth factor stimulus
negative regulation of pancreatic A cell differentiation
negative regulation of stem cell differentiation
negative regulation of pro-B cell differentiation
negative regulation of forebrain neuron differentiation
negative regulation of inner ear receptor cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
JUN kinase binding
protein complex binding
protein homodimerization activity
histone deacetylase binding
sequence-specific DNA binding
protein dimerization activity
chaperone binding
E-box binding
N-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0