Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPA8 SIRNA Assays ›
          Gene Symbol:
          Hspa8
          Gene Name:
          heat shock protein 8
          Gene Aliases:
          2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
          Chromosome Location:
          Chr.9: 40801273 - 40805199 on Build GRCm38
          UniGene ID:
          Mm.290774
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s67830

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hspa8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031165.4 NP_112442.2 5 1184
          GenBank mRNA AK004608.1 - 4 1473
          AK035286.1 - 5 1182
          AK075935.1 - 5 1181
          AK081701.1 - 5 1183
          AK098074.1 - 5 1181
          AK135281.1 - 1 353
          AK145416.1 - 5 1104
          AK145579.1 - 5 1178
          AK146547.1 - 5 1188
          AK146708.1 - 5 1179
          AK146985.1 - 5 1182
          AK147864.1 - 5 1182
          AK149954.1 - 5 1179
          AK150474.1 - 5 1176
          AK150498.1 - 5 1176
          AK150701.1 - 5 1186
          AK150958.1 - 5 1179
          AK151041.1 - 5 1182
          AK151065.1 - 5 1182
          AK151127.1 - 5 1185
          AK151287.1 - 5 1182
          AK151435.1 - 5 1182
          AK151516.1 - 5 1188
          AK151537.1 - 5 1179
          AK151741.1 - 5 1182
          AK151775.1 - 5 1184
          AK151808.1 - 5 1185
          AK151865.1 - 5 1182
          AK151892.1 - 5 1184
          AK151948.1 - 5 1185
          AK151997.1 - 5 1182
          AK152315.1 - 5 1185
          AK152598.1 - 5 1182
          AK152697.1 - 5 1182
          AK152703.1 - 5 1182
          AK152768.1 - 5 1185
          AK152803.1 - 5 1188
          AK153032.1 - 5 1184
          AK153834.1 - 5 1182
          AK157831.1 - 5 1182
          AK159479.1 - 5 1182
          AK162775.1 - 5 1182
          AK163987.1 - 5 1182
          AK164000.1 - 5 1182
          AK166545.1 - 5 1184
          AK166643.1 - 5 1196
          AK166721.1 - 5 1179
          AK166767.1 - 5 1182
          AK166776.1 - 5 1185
          AK166808.1 - 5 1182
          AK166830.1 - 5 1180
          AK166846.1 - 5 1182
          AK166861.1 - 5 1185
          AK166873.1 - 5 1182
          AK166908.1 - 5 1185
          AK166910.1 - 5 1182
          AK166913.1 - 5 1182
          AK166933.1 - 5 1182
          AK167043.1 - 5 1184
          AK167121.1 - 5 1185
          AK167122.1 - 5 1182
          AK167134.1 - 5 1180
          AK167163.1 - 5 1184
          AK167218.1 - 5 1182
          AK167229.1 - 5 1183
          AK167254.1 - 5 1183
          AK167414.1 - 5 1182
          AK167845.1 - 5 1185
          AK167910.1 - 5 1185
          AK168435.1 - 5 1185
          AK168492.1 - 5 1184
          AK168511.1 - 3 720
          AK168519.1 - 5 1182
          AK168542.1 - 5 1184
          AK168577.1 - 5 1182
          AK168625.1 - 5 1182
          AK168654.1 - 5 1182
          AK168711.1 - 5 1182
          AK168750.1 - 5 1185
          AK168776.1 - 5 1185
          AK168887.1 - 5 1184
          AK168934.1 - 5 1185
          AK169093.1 - 5 1182
          AK169179.1 - 5 1182
          AK169236.1 - 5 1182
          AK169293.1 - 5 1196
          AK169329.1 - 5 1185
          AK169594.1 - 5 1181
          AK171434.1 - 5 1183
          AK180175.1 - 1 89
          AK180949.1 - 1 209
          AK190071.1 - 1 210
          AK190740.1 - 1 209
          AK190962.1 - 1 209
          AK191872.1 - 1 209
          AK192391.1 - 1 209
          AK192614.1 - 1 209
          AK194175.1 - 1 209
          AK195094.1 - 1 209
          AK195617.1 - 1 210
          AK196257.1 - 1 209
          AK197810.1 - 1 209
          AK197951.1 - 1 209
          AK199432.1 - 1 209
          AK199584.1 - 1 209
          AK206309.1 - 1 209
          AK208890.1 - 1 99
          AK215442.1 - 1 209
          AK216947.1 - 1 209
          BC006722.1 - 5 1206 3711399
          BC066191.1 - 5 1138 30467052
          BC085486.1 - 5 1170 30540914
          BC089322.1 - 5 1163 30538640
          BC089457.1 - 5 1141 30628505
          BC094900.1 - 5 1098 30063483
          BC106169.1 - 3 862
          BC106193.1 - 5 1166 6329283
          M19141.1 - 5 1168
          U27129.1 - 5 1133

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_024351, XM_008764620, M11942, BC061547, FQ235115, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          protein folding
          RNA splicing
          protein refolding
          negative regulation of transcription, DNA-templated
          ATP metabolic process
          positive regulation of mRNA splicing, via spliceosome
          chaperone mediated protein folding requiring cofactor
          regulation of cell cycle
          chaperone-mediated protein folding
          clathrin coat disassembly
          negative regulation of fibril organization
          nucleotide binding
          G-protein coupled receptor binding
          receptor binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          MHC class II protein complex binding
          heat shock protein binding
          ubiquitin protein ligase binding
          ATPase activity, coupled
          ADP binding
          poly(A) RNA binding
          unfolded protein binding
          C3HC4-type RING finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0