Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HES1 SIRNA Assays ›
          Gene Symbol:
          HES1
          Gene Name:
          hes family bHLH transcription factor 1
          Gene Aliases:
          HES-1, HHL, HRY, bHLHb39
          Chromosome Location:
          Chr.3: 194136142 - 194138612 on Build GRCh38
          UniGene ID:
          Hs.250666
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s6920

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HES1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005524.3 NP_005515.1 2 364
          GenBank mRNA AF264785.1 - 2 125
          AK000415.1 - 2 361
          AK300482.1 - 2 355
          BC039152.1 - 2 312 4749611
          CR541843.1 - 2 125

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          liver development
          embryonic heart tube morphogenesis
          outflow tract morphogenesis
          regulation of secondary heart field cardioblast proliferation
          ventricular septum development
          transcription, DNA-templated
          protein complex assembly
          cell adhesion
          Notch signaling pathway
          smoothened signaling pathway
          STAT protein import into nucleus
          nervous system development
          positive regulation of cell proliferation
          cell migration
          telencephalon development
          midbrain-hindbrain boundary morphogenesis
          oculomotor nerve development
          trochlear nerve development
          hindbrain morphogenesis
          forebrain radial glial cell differentiation
          adenohypophysis development
          lung development
          positive regulation of BMP signaling pathway
          midbrain development
          pancreas development
          somatic stem cell population maintenance
          ascending aorta morphogenesis
          positive regulation of T cell proliferation
          positive regulation of tyrosine phosphorylation of Stat3 protein
          auditory receptor cell fate determination
          positive regulation of DNA binding
          regulation of fat cell differentiation
          negative regulation of auditory receptor cell differentiation
          positive regulation of Notch signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of mitotic cell cycle, embryonic
          lateral inhibition
          positive regulation of JAK-STAT cascade
          cell maturation
          thymus development
          cell morphogenesis involved in neuron differentiation
          positive regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          artery morphogenesis
          regulation of epithelial cell proliferation
          inner ear receptor stereocilium organization
          regulation of timing of neuron differentiation
          negative regulation of glial cell proliferation
          ventricular septum morphogenesis
          ureteric bud morphogenesis
          labyrinthine layer blood vessel development
          common bile duct development
          negative regulation of stomach neuroendocrine cell differentiation
          cardiac neural crest cell development involved in outflow tract morphogenesis
          pharyngeal arch artery morphogenesis
          glomerulus vasculature development
          comma-shaped body morphogenesis
          S-shaped body morphogenesis
          renal interstitial fibroblast development
          metanephric nephron tubule morphogenesis
          cochlea development
          establishment of epithelial cell polarity
          vascular smooth muscle cell development
          neuronal stem cell population maintenance
          positive regulation of protein targeting to mitochondrion
          negative regulation of pancreatic A cell differentiation
          negative regulation of stem cell differentiation
          negative regulation of pro-B cell differentiation
          negative regulation of forebrain neuron differentiation
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein homodimerization activity
          histone deacetylase binding
          sequence-specific DNA binding
          chaperone binding
          N-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0