Search
Search

Lrpap1| Gene Symbol: Lrpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013587.2 | NP_038615.2 | 4 | 498 | ||
| GenBank mRNA | AJ639614.1 | - | 4 | 505 | ||
| AK089250.1 | - | 4 | 501 | |||
| AK145710.1 | - | 4 | 541 | |||
| AK150451.1 | - | 4 | 512 | |||
| AK152485.1 | - | 4 | 512 | |||
| AK153323.1 | - | 4 | 512 | |||
| AK166616.1 | - | 4 | 501 | |||
| AK166746.1 | - | 4 | 512 | |||
| AK166819.1 | - | 4 | 516 | |||
| AK168421.1 | - | 4 | 501 | |||
| AK169132.1 | - | 4 | 506 | |||
| AK171471.1 | - | 4 | 501 | |||
| BC032170.1 | - | 4 | 501 | |||
| BC046641.1 | - | 4 | 490 | |||
| BC057979.1 | - | 4 | 469 | |||
| BC059887.1 | - | 4 | 315 | |||
| BC094324.1 | - | 4 | 498 | 6402163 | ||
| D00622.1 | - | 4 | 491 | |||
| S67967.1 | - | 4 | 524 | |||