Search
Search

Tpsb2| Gene Symbol: Tpsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010781.3 | NP_034911.3 | 5 | 796 | ||
| GenBank mRNA | BC024374.1 | - | 5 | 796 | 4983727 | |
| L31853.1 | - | 5 | 790 | |||
| M57625.1 | - | 3052 | ||||
| M57626.1 | - | 6 | 818 | |||