Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEN1 SIRNA Assays ›
          Gene Symbol:
          Men1
          Gene Name:
          multiple endocrine neoplasia 1
          Gene Aliases:
          AW045611
          Chromosome Location:
          Chr.19: 6334979 - 6340894 on Build GRCm38
          UniGene ID:
          Mm.12917
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s69786

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Men1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001168488.1 NP_001161960.1 6, 7 1029
          NM_001168489.1 NP_001161961.1 6, 7 1007
          NM_001168490.1 NP_001161962.1 5, 6 1343
          NM_008583.2 NP_032609.1 5, 6 1343
          GenBank mRNA AB023401.1 - 5, 6 970
          AF016398.1 - 6, 7 982
          AF072755.1 - 6, 7 966
          AF109389.1 - 5, 6 1342
          AF130368.1 - 5, 6 1325
          AK133984.1 - 6, 7 966
          AK154371.1 - 6, 7 988
          AK164700.1 - 6, 7 964
          AK170713.1 - 5, 6 1323
          BC006610.1 - 6, 7 962
          BC036287.1 - 5, 6 1271 4189611

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          ossification
          leukocyte homeostasis
          negative regulation of protein phosphorylation
          osteoblast fate commitment
          osteoblast development
          type B pancreatic cell differentiation
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          cell cycle arrest
          negative regulation of cell proliferation
          response to UV
          embryo development
          response to gamma radiation
          negative regulation of cell-substrate adhesion
          chromatin modification
          histone methylation
          hemopoiesis
          positive regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of histone methylation
          positive regulation of protein binding
          regulation of activin receptor signaling pathway
          histone lysine methylation
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of cell differentiation
          negative regulation of osteoblast differentiation
          positive regulation of osteoblast differentiation
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of JNK cascade
          negative regulation of organ growth
          embryonic skeletal system morphogenesis
          negative regulation of epithelial cell proliferation
          positive regulation of cell division
          negative regulation of telomerase activity
          palate development
          maternal process involved in female pregnancy
          regulation of type B pancreatic cell proliferation
          response to transforming growth factor beta
          negative regulation of cell cycle G1/S phase transition
          four-way junction DNA binding
          Y-form DNA binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          protein binding
          histone-lysine N-methyltransferase activity
          protein binding, bridging
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein N-terminus binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0