Search
Search

Mpv17| Gene Symbol: Mpv17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294322.1 | NP_001281251.1 | 4 | 273 | ||
| NM_001294324.1 | NP_001281253.1 | 4 | 302 | |||
| NM_008622.6 | NP_032648.1 | 4 | 273 | |||
| GenBank mRNA | AK040477.1 | - | 4 | 243 | ||
| BC013452.1 | - | 3 | 226 | 4237284 | ||
| BY258057.1 | - | 4 | 244 | |||
| CK622051.1 | - | 3 | 215 | |||
| DV062332.1 | - | 4 | 355 | |||
| DV660230.1 | - | 2 | 90 | |||
| M36411.1 | - | 4 | 255 | |||