Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYH9 SIRNA Assays ›
          Gene Symbol:
          Myh9
          Gene Name:
          myosin, heavy polypeptide 9, non-muscle
          Gene Aliases:
          Fltn, Myhn-1, Myhn1, NMHCIIA, NMMHC-A, NMMHC-IIA, TU72.6
          Chromosome Location:
          Chr.15: 77760585 - 77842175 on Build GRCm38
          UniGene ID:
          Mm.29677
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s70268

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Myh9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022410.3 NP_071855.2 3 621
          GenBank mRNA AJ312390.1 - 3 545
          AK040822.1 - 3 552
          AK053221.1 - 3 557
          AK083071.1 - 3 551
          AK147203.1 - 3 544
          AK147206.1 - 3 553
          AK147208.1 - 3 561
          AK147209.1 - 3 552
          AK147210.1 - 3 552
          AK147211.1 - 3 596
          AK147215.1 - 3 539
          AK147216.1 - 3 494
          AK147221.1 - 3 499
          AK147222.1 - 3 540
          AK147223.1 - 3 545
          AK147233.1 - 3 552
          AK147235.1 - 3 552
          AK147296.1 - 3 552
          AK147407.1 - 3 555
          AK147430.1 - 3 562
          AK165379.1 - 3 502
          BC116417.1 - 3 506
          BC116418.1 - 3 506
          BC128497.1 - 3 506
          BC128498.1 - 3 506

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic spindle organization
          cell morphogenesis involved in differentiation
          cytokinesis
          angiogenesis
          in utero embryonic development
          establishment of T cell polarity
          membrane protein ectodomain proteolysis
          phagocytosis, engulfment
          movement of cell or subcellular component
          cell adhesion
          myoblast fusion
          regulation of cell shape
          protein transport
          single organismal cell-cell adhesion
          actin filament polymerization
          actin filament-based movement
          platelet formation
          actomyosin structure organization
          actin cytoskeleton reorganization
          uropod organization
          blood vessel endothelial cell migration
          establishment of meiotic spindle localization
          actin filament capping
          platelet aggregation
          negative regulation of actin filament severing
          positive regulation of protein processing in phagocytic vesicle
          microfilament motor activity
          nucleotide binding
          motor activity
          actin binding
          protein binding
          calmodulin binding
          ATP binding
          ATPase activity
          protein domain specific binding
          actin-dependent ATPase activity
          protein homodimerization activity
          protein anchor
          ADP binding
          poly(A) RNA binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0