Search
Search

Nfe2l1| Gene Symbol: Nfe2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130450.1 | NP_001123922.1 | 2 | 828 | ||
| NM_008686.3 | NP_032712.2 | 2 | 886 | |||
| GenBank mRNA | AF071084.1 | - | 1 | 232 | ||
| AK131183.1 | - | 2 | 821 | |||
| AK152040.1 | - | 2 | 867 | |||
| AK153529.1 | - | 2 | 868 | |||
| AK154167.1 | - | 2 | 797 | |||
| BC043063.1 | - | 1 | 468 | 5705669 | ||
| BC047283.1 | - | 1 | 228 | 6489220 | ||
| BC099895.1 | - | 2 | 852 | |||
| X78709.1 | - | 1 | 264 | |||