Search
Search

Nxn| Gene Symbol: Nxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008750.5 | NP_032776.1 | 5 | 890 | ||
| GenBank mRNA | AB095441.1 | - | 5 | 735 | ||
| AB095442.1 | - | 5 | 735 | |||
| AB095443.1 | - | 5 | 735 | |||
| AB095444.1 | - | 5 | 735 | |||
| AK012352.1 | - | 5 | 875 | |||
| AK048109.1 | - | 5 | 890 | |||
| AK149075.1 | - | 5 | 874 | |||
| AK192350.1 | - | 1 | 26 | |||
| BC004688.1 | - | 5 | 850 | 3501314 | ||
| BC047339.1 | - | 4 | 422 | |||
| BC058244.1 | - | 5 | 890 | 6312910 | ||