Search
Search

ID3| Gene Symbol: ID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002167.4 | NP_002158.3 | 3 | 1130 | ||
| GenBank mRNA | A17546.1 | - | 3 | 884 | ||
| AK225965.1 | - | 1 | 1491 | |||
| BC003107.1 | - | 3 | 848 | 3543936 | ||
| X66924.1 | - | 3 | 884 | |||
| X69111.1 | - | 3 | 1129 | |||