Search
Search

IDH1| Gene Symbol: IDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282386.1 | NP_001269315.1 | 9 | 1351 | ||
| NM_001282387.1 | NP_001269316.1 | 9 | 1508 | |||
| NM_005896.3 | NP_005887.2 | 9 | 1440 | |||
| GenBank mRNA | AF113917.1 | - | 9 | 1331 | ||
| AK302105.1 | - | 7 | 886 | |||
| AK312242.1 | - | 9 | 1411 | |||
| AL136702.1 | - | 9 | 1372 | |||
| BC012846.1 | - | 9 | 1391 | 3880331 | ||
| BC021046.1 | - | 9 | 1337 | |||
| BC093020.1 | - | 9 | 1369 | 30528912 | ||
| BX537411.1 | - | 9 | 1340 | |||
| CR533522.1 | - | 7 | 1145 | |||
| CR536499.1 | - | 7 | 1145 | |||
| CR541695.1 | - | 7 | 1145 | |||
| DQ403082.1 | - | 7 | 1058 | |||
| EU794623.1 | - | 9 | 1379 | |||
| FJ224342.1 | - | 9 | 1340 | |||