Search
Search

Pck1| Gene Symbol: Pck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011044.2 | NP_035174.1 | 5 | 847 | ||
| GenBank mRNA | AK028046.1 | - | 5 | 848 | ||
| AK030327.1 | - | 5 | 847 | |||
| AK133496.1 | - | 5 | 848 | |||
| AK149525.1 | - | 5 | 848 | |||
| BC028342.1 | - | 5 | 849 | |||
| BC037629.1 | - | 5 | 842 | 4973187 | ||