Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLCB1 SIRNA Assays ›
          Gene Symbol:
          Plcb1
          Gene Name:
          phospholipase C, beta 1
          Gene Aliases:
          3110043I21Rik, AI132408, Plcb, mKIAA0581
          Chromosome Location:
          Chr.2: 134786164 - 135475258 on Build GRCm38
          UniGene ID:
          Mm.330607
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s71793

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Plcb1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145830.1 NP_001139302.1 26 3103
          NM_019677.2 NP_062651.2 26 3103
          GenBank mRNA AF498249.1 - 26 2734
          AK014170.1 - 4 338
          AK129166.1 - 18 1968
          AK141164.1 - 26 3051
          BC058710.1 - 26 3070 6842016
          U85712.1 - 26 2737
          U85713.1 - 26 2737
          U85714.1 - 26 2737

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          oocyte maturation
          mRNA processing
          lipid metabolic process
          cell adhesion
          signal transduction
          G-protein coupled acetylcholine receptor signaling pathway
          glutamate receptor signaling pathway
          brain development
          memory
          regulation of G-protein coupled receptor protein signaling pathway
          insulin receptor signaling pathway
          lipid catabolic process
          cerebral cortex development
          erythrocyte differentiation
          macrophage differentiation
          positive regulation of sodium:proton antiporter activity
          positive regulation of interleukin-12 production
          response to monosaccharide
          intracellular signal transduction
          interleukin-12-mediated signaling pathway
          interleukin-15-mediated signaling pathway
          positive regulation of embryonic development
          response to peptide hormone
          positive regulation of GTPase activity
          fat cell differentiation
          positive regulation of myoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of JNK cascade
          insulin-like growth factor receptor signaling pathway
          positive regulation of developmental growth
          regulation of cell cycle
          activation of meiosis involved in egg activation
          interleukin-1-mediated signaling pathway
          regulation of fertilization
          positive regulation of G1/S transition of mitotic cell cycle
          positive regulation of acrosome reaction
          negative regulation of monocyte extravasation
          positive regulation of CD24 biosynthetic process
          phosphatidylinositol phospholipase C activity
          signal transducer activity
          GTPase activator activity
          calcium ion binding
          protein binding
          calmodulin binding
          lamin binding
          phosphatidylinositol-4,5-bisphosphate binding
          phosphoric diester hydrolase activity
          hydrolase activity
          enzyme binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline