Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL4 SIRNA Assays ›
          Gene Symbol:
          IL4
          Gene Name:
          interleukin 4
          Gene Aliases:
          BCGF-1, BCGF1, BSF-1, BSF1, IL-4
          Chromosome Location:
          Chr.5: 132673986 - 132682678 on Build GRCh38
          UniGene ID:
          Hs.73917
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7301

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000589.3 NP_000580.1 3 377
          NM_172348.2 NP_758858.1 2 329
          GenBank mRNA AB102862.1 - 2 264
          AF043336.1 - 2 195
          BC066278.1 - 3 377
          BC067514.1 - 3 377 6971780
          BC067515.1 - 3 377 6971782
          BC070123.1 - 3 374 30415895
          CD640452.1 - 3 377
          EF173851.1 - 2 182
          JX878389.1 - 4 413
          M13982.1 - 3 375
          X81851.1 - 2 264

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          innate immune response in mucosa
          positive regulation of defense response to virus by host
          T-helper 1 cell lineage commitment
          negative regulation of acute inflammatory response
          negative regulation of chronic inflammatory response
          chemotaxis
          immune response
          cellular defense response
          female pregnancy
          response to nutrient
          cholesterol metabolic process
          regulation of proton transport
          negative regulation of epithelial cell migration
          response to organic cyclic compound
          B cell differentiation
          positive regulation of B cell proliferation
          B cell costimulation
          positive regulation of interleukin-10 production
          positive regulation of interleukin-13 production
          response to cytokine
          T-helper 2 cell cytokine production
          type 2 immune response
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          regulation of phosphorylation
          response to drug
          positive regulation of tyrosine phosphorylation of Stat5 protein
          defense response to protozoan
          myeloid dendritic cell differentiation
          negative regulation of macrophage activation
          negative regulation of apoptotic process
          positive regulation of mast cell degranulation
          negative regulation of nitric oxide biosynthetic process
          T-helper 2 cell differentiation
          positive regulation of chemokine biosynthetic process
          connective tissue growth factor biosynthetic process
          regulation of isotype switching
          positive regulation of MHC class II biosynthetic process
          response to ethanol
          positive regulation of T cell differentiation
          negative regulation of osteoclast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of isotype switching to IgE isotypes
          positive regulation of isotype switching to IgG isotypes
          regulation of immune response
          positive regulation of sequence-specific DNA binding transcription factor activity
          retina development in camera-type eye
          negative regulation of white fat cell proliferation
          cellular response to mercury ion
          positive regulation of mononuclear cell migration
          dendritic cell differentiation
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of myoblast fusion
          negative regulation of complement-dependent cytotoxicity
          negative regulation of T-helper 17 cell differentiation
          negative regulation of endothelial cell apoptotic process
          positive regulation of eosinophil chemotaxis
          negative regulation of extrinsic apoptotic signaling pathway
          cytokine activity
          interleukin-4 receptor binding
          protein binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0