Search
Search

KIF5B| Gene Symbol: KIF5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004521.2 | NP_004512.1 | 20, 21 | 2785 | ||
| XM_017016224.1 | XP_016871713.1 | 20, 21 | 2391 | |||
| GenBank mRNA | AK289413.1 | - | 20, 21 | 2731 | ||
| AK299542.1 | - | 1156 | ||||
| BC126279.1 | - | 20, 21 | 2538 | 8991995 | ||
| BC126281.1 | - | 20, 21 | 2538 | 8991997 | ||
| FJ224324.1 | - | 20, 21 | 2628 | |||
| X65873.1 | - | 20, 21 | 2628 | |||