Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMARCA4 SIRNA Assays ›
          Gene Symbol:
          Smarca4
          Gene Name:
          SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
          Gene Aliases:
          Brg1, HP1-BP72, SNF2beta, SW1/SNF, b2b508.1Clo, b2b692Clo
          Chromosome Location:
          Chr.9: 21616106 - 21704230 on Build GRCm38
          UniGene ID:
          Mm.286593
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s74000

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Smarca4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001174078.1 NP_001167549.1 24, 25 3642
          NM_001174079.1 NP_001167550.1 24, 25 3642
          NM_011417.3 NP_035547.2 24, 25 3642
          GenBank mRNA AK141517.1 - 21, 22 2796
          AK147047.1 - 9, 10 958
          AK147285.1 - 24, 25 3596
          AK160825.1 - 24, 25 3546
          AK166837.1 - 24, 25 3547
          BC023186.1 - 3, 4 231
          BC026672.1 - 7, 8 791
          BC060229.1 - 24, 25 3536
          BC079560.1 - 24, 25 3642 30533489
          S68108.1 - 14, 15 1608

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          blood vessel development
          vasculogenesis
          in utero embryonic development
          blastocyst growth
          blastocyst hatching
          liver development
          outflow tract morphogenesis
          nucleosome assembly
          nucleosome disassembly
          chromatin remodeling
          methylation-dependent chromatin silencing
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          nervous system development
          glial cell fate determination
          heart development
          positive regulation of cell proliferation
          DNA methylation on cytosine within a CG sequence
          chromatin modification
          stem cell population maintenance
          neurogenesis
          positive regulation of Wnt signaling pathway
          extracellular matrix organization
          keratinocyte differentiation
          negative regulation of cell growth
          regulation of cell migration
          forebrain development
          hindbrain development
          embryonic hindlimb morphogenesis
          aortic smooth muscle cell differentiation
          ATP-dependent chromatin remodeling
          negative regulation of apoptotic process
          positive regulation of DNA binding
          positive regulation by host of viral transcription
          histone H3 acetylation
          positive regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic organ morphogenesis
          epidermis morphogenesis
          positive regulation of sequence-specific DNA binding transcription factor activity
          definitive erythrocyte differentiation
          heart trabecula formation
          negative regulation of androgen receptor signaling pathway
          pharyngeal arch artery morphogenesis
          lens fiber cell development
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          nucleotide binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription coactivator activity
          p53 binding
          DNA binding
          chromatin binding
          transcription coactivator activity
          transcription corepressor activity
          helicase activity
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          transcription factor binding
          hydrolase activity
          hydrolase activity, acting on acid anhydrides
          hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
          ATPase activity
          Tat protein binding
          nucleosomal DNA binding
          protein complex binding
          histone binding
          protein N-terminus binding
          androgen receptor binding
          DNA polymerase binding
          lysine-acetylated histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0