Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INPP4A SIRNA Assays ›
          Gene Symbol:
          INPP4A
          Gene Name:
          inositol polyphosphate-4-phosphatase type I A
          Gene Aliases:
          INPP4, TVAS1
          Chromosome Location:
          Chr.2: 98444592 - 98594068 on Build GRCh38
          UniGene ID:
          Hs.469386
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7450
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INPP4A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134224.1 NP_001127696.1 23 2843
          NM_001134225.1 NP_001127697.1 22 2828
          NM_001566.2 NP_001557.1 23 2726
          NM_004027.2 NP_004018.1 23 2726
          XM_006712499.3 XP_006712562.1 23 2841
          XM_006712500.3 XP_006712563.1 23 2838
          XM_006712501.3 XP_006712564.1 22 2823
          XM_006712502.3 XP_006712565.1 22 2808
          XM_006712503.3 XP_006712566.1 23 2841
          XM_006712504.3 XP_006712567.1 23 2724
          XM_006712505.3 XP_006712568.1 23 2841
          XM_006712506.3 XP_006712569.1 22 2691
          XM_006712507.3 XP_006712570.1 23 2724
          XM_006712508.3 XP_006712571.1 23 2841
          XM_011511122.2 XP_011509424.1 22 2791
          XM_011511123.2 XP_011509425.1 23 2841
          XM_011511124.2 XP_011509426.1 22 2706
          XM_011511125.2 XP_011509427.1 20 2723
          XM_011511127.2 XP_011509429.1 11 1526
          XM_017003993.1 XP_016859482.1 23 2565
          XM_017003994.1 XP_016859483.1 22 2706
          XM_017003995.1 XP_016859484.1 22 2688
          XM_017003996.1 XP_016859485.1 23 2841
          XM_017003997.1 XP_016859486.1 23 2783
          XM_017003998.1 XP_016859487.1 23 2780
          XM_017003999.1 XP_016859488.1 22 2764
          XM_017004000.1 XP_016859489.1 22 2691
          XM_017004001.1 XP_016859490.1 21 2673
          XM_017004002.1 XP_016859491.1 21 2591
          XM_017004003.1 XP_016859492.1 23 2724
          GenBank mRNA AF368319.1 - 21 2450
          BC028361.2 - 22 2828 4830663
          U26398.1 - 23 2627
          U96919.1 - 23 2627

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00244467_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylinositol biosynthetic process
          signal transduction
          dephosphorylation
          phosphatidylinositol-3-phosphate biosynthetic process
          inositol phosphate metabolic process
          protein binding
          phosphatidylinositol-3,4-bisphosphate 4-phosphatase activity
          inositol-1,3,4-trisphosphate 4-phosphatase activity
          phosphatidylinositol-4,5-bisphosphate 4-phosphatase activity
          inositol-3,4-bisphosphate 4-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0