Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRF3 SIRNA Assays ›
Gene Symbol:
IRF3
Gene Name:
interferon regulatory factor 3
Gene Aliases:
IIAE7
Chromosome Location:
Chr.19: 49659569 - 49665875 on Build GRCh38
UniGene ID:
Hs.289052
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s7507
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRF3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197122.1
NP_001184051.1
4
623
NM_001197123.1
NP_001184052.1
4
681
NM_001197124.1
NP_001184053.1
4
623
NM_001197125.1
NP_001184054.1
3
450
NM_001197126.1
NP_001184055.1
2
278
NM_001197127.1
NP_001184056.1
3
450
NM_001197128.1
NP_001184057.1
2
278
NM_001571.5
NP_001562.1
4
623
NR_045568.1
4
770
XM_006723197.1
XP_006723260.1
4
601
XM_006723198.1
XP_006723261.1
4
537
XM_006723200.1
XP_006723263.1
3
286
XM_006723201.1
XP_006723264.1
3
359
XM_006723202.2
XP_006723265.1
4
508
XM_017026766.1
XP_016882255.1
4
592
XM_017026767.1
XP_016882256.1
4
537
GenBank mRNA
AB102884.1
-
2
198
AB102887.1
-
3
370
AK057577.1
-
3
1389
AK292027.1
-
4
752
AK314421.1
-
4
536
BC000660.2
-
4
525
3347928
BC009395.2
-
4
550
4135383
BC071721.1
-
3
712
5494536
BP347627.1
-
3
291
CB961512.1
-
4
631
HM357932.1
-
4
511
HM357933.1
-
2
108
HM357934.1
-
2
108
HM357935.1
-
3
280
HM357936.1
-
4
453
HM357937.1
-
3
280
Z56281.1
-
4
416
Back To Top
More Information
Panther Classification:
Molecular Function -
winged helix/forkhead transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
apoptotic process
cellular response to DNA damage stimulus
lipopolysaccharide-mediated signaling pathway
negative regulation of type I interferon production
positive regulation of type I interferon production
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
TRIF-dependent toll-like receptor signaling pathway
MDA-5 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to exogenous dsRNA
type I interferon biosynthetic process
positive regulation of cytokine secretion
defense response to virus
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
positive regulation of type I interferon-mediated signaling pathway
cellular response to dsRNA
macrophage apoptotic process
programmed necrotic cell death
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription cofactor activity
protein binding
identical protein binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0