Search
Search

ITGB2| Gene Symbol: ITGB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000211.4 | NP_000202.3 | 6 | 874 | ||
| NM_001127491.2 | NP_001120963.2 | 6 | 864 | |||
| NM_001303238.1 | NP_001290167.1 | 6 | 829 | |||
| XM_006724001.1 | XP_006724064.1 | 6 | 891 | |||
| XM_017028341.1 | XP_016883830.1 | 6 | 886 | |||
| GenBank mRNA | AB208909.1 | - | 6 | 784 | ||
| AK095992.1 | - | 6 | 829 | |||
| AK128344.1 | - | 6 | 699 | |||
| AK222505.1 | - | 6 | 731 | |||
| AK303488.1 | - | 6 | 742 | |||
| BC005861.2 | - | 5 | 685 | 3532902 | ||
| BC021077.1 | - | 5 | 608 | |||
| L78790.1 | - | 5 | 522 | |||
| M15395.1 | - | 6 | 713 | |||
| Y00057.1 | - | 5 | 617 | |||