Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNT3A SIRNA Assays ›
Gene Symbol:
Wnt3a
Gene Name:
wingless-type MMTV integration site family, member 3A
Gene Aliases:
Wnt-3a, vt
Chromosome Location:
Chr.11: 59248036 - 59290751 on Build GRCm38
UniGene ID:
Mm.1367
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s76082
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Wnt3a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009522.2
NP_033548.1
4
945
GenBank mRNA
X56842.1
-
4
969
Back To Top
More Information
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
in utero embryonic development
somitogenesis
positive regulation of cytokine production
positive regulation of protein phosphorylation
heart looping
positive regulation of receptor internalization
negative regulation of heart induction by canonical Wnt signaling pathway
signal transduction
cell-cell signaling
multicellular organism development
determination of left/right symmetry
axonogenesis
axon guidance
mesoderm development
positive regulation of cell proliferation
organ morphogenesis
anterior/posterior pattern specification
COP9 signalosome assembly
negative regulation of neuron projection development
Wnt signaling pathway
spinal cord association neuron differentiation
hippocampus development
cell proliferation in forebrain
Wnt signaling pathway involved in forebrain neuroblast division
dorsal/ventral neural tube patterning
hemopoiesis
platelet activation
extracellular matrix organization
BMP signaling pathway
mammary gland development
positive regulation of B cell proliferation
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
cellular protein localization
positive regulation of catenin import into nucleus
skeletal muscle cell differentiation
post-anal tail morphogenesis
inner ear morphogenesis
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of cell differentiation
negative regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
somatic stem cell division
positive regulation of mesodermal cell fate specification
paraxial mesodermal cell fate commitment
positive regulation of collateral sprouting in absence of injury
negative regulation of axon extension involved in axon guidance
negative regulation of neurogenesis
regulation of axonogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
palate development
canonical Wnt signaling pathway
cardiac muscle cell fate commitment
positive regulation of protein tyrosine kinase activity
positive regulation of dermatome development
canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
regulation of microtubule cytoskeleton organization
platelet aggregation
axis elongation involved in somitogenesis
positive regulation of canonical Wnt signaling pathway
positive regulation of cell-cell adhesion mediated by cadherin
positive regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
positive regulation of neural precursor cell proliferation
positive regulation of receptor activity
positive regulation of cardiac muscle cell differentiation
transcription coactivator activity
receptor binding
frizzled binding
frizzled-2 binding
protein binding
protein domain specific binding
receptor agonist activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-jct7h:80/100.66.72.173:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0