Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT7B SIRNA Assays ›
          Gene Symbol:
          Wnt7b
          Gene Name:
          wingless-type MMTV integration site family, member 7B
          Gene Aliases:
          Wnt-7b
          Chromosome Location:
          Chr.15: 85535437 - 85581821 on Build GRCm38
          UniGene ID:
          Mm.306946
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s76099

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Wnt7b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163633.1 NP_001157105.1 3 896
          NM_001163634.1 NP_001157106.1 3 980
          NM_009528.3 NP_033554.3 3 859
          GenBank mRNA AK087499.1 - 3 897
          BC052018.1 - 3 620 6401357
          BC058398.1 - 3 855
          BC066003.1 - 3 980 5701652
          M89802.1 - 3 488

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          in utero embryonic development
          vasculature development
          metanephros morphogenesis
          signal transduction
          activation of JUN kinase activity
          multicellular organism development
          positive regulation of cell proliferation
          positive regulation of gene expression
          Wnt signaling pathway
          establishment or maintenance of polarity of embryonic epithelium
          cell proliferation in forebrain
          central nervous system vasculogenesis
          neuron differentiation
          lung development
          neuron projection development
          oxygen homeostasis
          regulation of cell projection size
          odontogenesis of dentin-containing tooth
          homeostatic process
          cellular metabolic process
          cell fate commitment
          positive regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          positive regulation of JNK cascade
          embryonic organ development
          neuron projection morphogenesis
          stem cell development
          negative regulation of neurogenesis
          synapse organization
          smooth muscle cell differentiation
          anatomical structure regression
          canonical Wnt signaling pathway
          lung morphogenesis
          lung epithelium development
          lobar bronchus development
          lung-associated mesenchyme development
          trachea cartilage morphogenesis
          developmental growth involved in morphogenesis
          embryonic placenta morphogenesis
          chorio-allantoic fusion
          renal inner medulla development
          renal outer medulla development
          outer medullary collecting duct development
          inner medullary collecting duct development
          metanephric collecting duct development
          metanephric epithelium development
          metanephric loop of Henle development
          apoptotic process involved in patterning of blood vessels
          positive regulation of receptor activity
          receptor binding
          frizzled binding
          protein binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0