Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002221.3 | NP_002212.3 | 6 | 2870 | ||
GenBank mRNA | AJ242780.1 | - | 5 | 2587 | ||
AK025304.1 | - | 1 | 1875 | |||
AK289700.1 | - | 5 | 1316 | |||
AK313801.1 | - | 5 | 1335 | |||
AY513281.1 | - | 6 | 1111 | |||
X57206.1 | - | 5 | 1209 | |||
Y18024.2 | - | 5 | 2530 |