Search
Search

Sh3yl1| Gene Symbol: Sh3yl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289480.1 | NP_001276409.1 | 3, 4 | 736 | ||
| NM_013709.5 | NP_038737.1 | 4, 5 | 850 | |||
| GenBank mRNA | AK141199.1 | - | 4, 5 | 851 | ||
| AK168483.1 | - | 5, 6 | 478 | |||
| BC010296.1 | - | 3, 4 | 298 | 3154966 | ||
| BC058557.1 | - | 3, 4 | 713 | 6313231 | ||
| BC061488.1 | - | 4, 5 | 790 | 4483634 | ||
| BC110043.1 | - | 4, 5 | 828 | 5149633 | ||
| D85926.1 | - | 4, 5 | 333 | |||