Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNMA1 SIRNA Assays ›
          Gene Symbol:
          KCNMA1
          Gene Name:
          potassium calcium-activated channel subfamily M alpha 1
          Gene Aliases:
          BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
          Chromosome Location:
          Chr.10: 76869601 - 77637819 on Build GRCh38
          UniGene ID:
          Hs.144795
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7787

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KCNMA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001014797.2 NP_001014797.1 23 2870
          NM_001161352.1 NP_001154824.1 23 3032
          NM_001161353.1 NP_001154825.1 23 2981
          NM_001271518.1 NP_001258447.1 22 2708
          NM_001271519.1 NP_001258448.1 23 2867
          NM_001322829.1 NP_001309758.1 23 2867
          NM_001322830.1 NP_001309759.1 24 2879
          NM_001322832.1 NP_001309761.1 22 2858
          NM_001322835.1 NP_001309764.1 23 2870
          NM_001322836.1 NP_001309765.1 23 2867
          NM_001322837.1 NP_001309766.1 23 2870
          NM_001322838.1 NP_001309767.1 24 2905
          NM_002247.3 NP_002238.2 22 2858
          XM_005269776.3 XP_005269833.1 22 2847
          XM_005269778.1 XP_005269835.1 24 3033
          XM_005269781.2 XP_005269838.1 23 2953
          XM_005269787.3 XP_005269844.1 22 2847
          XM_005269789.2 XP_005269846.1 23 2791
          XM_005269792.2 XP_005269849.1 23 2866
          XM_005269796.2 XP_005269853.1 22 2779
          XM_006717826.2 XP_006717889.1 22 2779
          XM_011539773.1 XP_011538075.1 25 3165
          XM_011539774.1 XP_011538076.1 25 3078
          XM_011539775.1 XP_011538077.1 24 3153
          XM_011539776.1 XP_011538078.1 24 3153
          XM_011539777.1 XP_011538079.1 24 3033
          XM_011539778.2 XP_011538080.1 23 2953
          XM_011539779.1 XP_011538081.1 24 3066
          XM_011539780.1 XP_011538082.1 24 3033
          XM_011539781.2 XP_011538083.1 24 2991
          XM_011539782.2 XP_011538084.1 23 2931
          XM_011539783.1 XP_011538085.1 24 2946
          XM_011539785.1 XP_011538087.1 23 2856
          XM_017016207.1 XP_016871696.1 25 3165
          XM_017016208.1 XP_016871697.1 25 3165
          XM_017016209.1 XP_016871698.1 24 3153
          XM_017016210.1 XP_016871699.1 25 3165
          XM_017016211.1 XP_016871700.1 25 3078
          XM_017016212.1 XP_016871701.1 24 2991
          XM_017016213.1 XP_016871702.1 24 3066
          XM_017016214.1 XP_016871703.1 23 2979
          XM_017016215.1 XP_016871704.1 24 2946
          XM_017016216.1 XP_016871705.1 24 2991
          XM_017016217.1 XP_016871706.1 23 2931
          XM_017016218.1 XP_016871707.1 22 2316
          XM_017016222.1 XP_016871711.1 22 2847
          XM_017016223.1 XP_016871712.1 21 2243
          GenBank mRNA AB113382.1 - 23 2797
          AB113575.1 - 23 2732
          AB209488.1 - 22 2277
          AB524033.1 - 22 2531
          AF025999.1 - 22 2609
          AF118141.1 - 22 2486
          AK310379.1 - 24 2905
          AY040849.1 - 22 2623
          BC137115.1 - 22 2761 9021112
          BC137137.1 - 22 2761 9021134
          BC144496.1 - 23 2770 9053023
          BX648925.1 - 19 2023
          U02632.1 - 16 1803
          U11058.2 - 22 2572
          U11717.1 - 22 2633
          U13913.1 - 22 2759
          U23767.1 - 22 2681

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031828, AY344964, AY330292, AF135265, U40603, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001253365, NM_001253366, NM_001253378, NM_001253377, NM_001253367, and others.

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          potassium ion transport
          response to osmotic stress
          cellular potassium ion homeostasis
          response to carbon monoxide
          regulation of ion transmembrane transport
          regulation of membrane potential
          positive regulation of apoptotic process
          negative regulation of cell volume
          response to calcium ion
          micturition
          smooth muscle contraction involved in micturition
          potassium ion transmembrane transport
          actin binding
          voltage-gated potassium channel activity
          protein binding
          calcium-activated potassium channel activity
          metal ion binding
          large conductance calcium-activated potassium channel activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0